The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003137.4 | NP_003128.3 | 2 - 3 | 200 | 56 |
GenBank mRNA | AJ318054.1 | - | 1 - 2 | 598 | 56 |
AK292277.1 | - | 2 - 3 | 145 | 56 | |
AK312712.1 | - | 2 - 3 | 92 | 56 | |
BC038292.1 | - | 2 - 3 | 165 | 56 | |
BG772464.1 | - | 2 - 3 | 208 | 56 | |
BX648663.1 | - | 2 - 3 | 117 | 56 | |
BX648697.1 | - | 2 - 3 | 118 | 56 | |
DB233331.1 | - | 2 - 3 | 200 | 56 |