Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNH2
Entrez Gene ID
3757
Gene Name
potassium voltage-gated channel subfamily H member 2
Gene Aliases
ERG-1, ERG1, H-ERG, HERG, HERG1, Kv11.1, LQT2, SQT1
Chromosome Location
Chr.7: 150944956 - 150978314 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000238.3 NP_000229.1 14 - 15 3730 60
NM_172057.2 NP_742054.1 10 - 11 2635 60
XM_011516185.2 XP_011514487.1 13 - 14 3141 60
XM_017012195.1 XP_016867684.1 13 - 14 3811 60
XM_017012196.1 XP_016867685.1 14 - 15 3271 60
GenBank mRNA AF363636.1 - 14 - 15 3329 60
AJ512214.1 - 10 - 11 2326 60
BC004311.2 - 6 - 7 1184 60
BC127673.1 - 5 - 6 1473 60
BM674996.1 - 2 - 3 60
DQ525913.1 - 14 - 15 3730 60
FJ938021.1 - 12 - 13 4162 60
U04270.1 - 14 - 15 3512 60

View Full Product Details