The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030876.5 | NP_110503.3 | 1 - 1 | 936 | 106 |
XM_011514929.2 | XP_011513231.1 | 3 - 3 | 1111 | 106 | |
XM_011514930.2 | XP_011513232.1 | 2 - 2 | 989 | 106 | |
GenBank mRNA | AJ459859.1 | - | 1 - 1 | 939 | 106 |
BC069149.1 | - | 1 - 1 | 854 | 106 |