The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_206914.1 | NP_996797.1 | 2 - 3 | 415 | 97 |
GenBank mRNA | AL050100.1 | - | 2 - 3 | 337 | 97 |