The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003630.2 | NP_003621.1 | 10 - 11 | 1207 | 105 |
GenBank mRNA | AB035307.1 | - | 10 - 11 | 1091 | 105 |
AJ001625.1 | - | 10 - 11 | 1008 | 105 | |
AJ131389.1 | - | 10 - 11 | 1004 | 105 | |
AK025644.1 | - | - | 1073 | 105 | |
AY277600.1 | - | 10 - 11 | 964 | 105 | |
BC014551.1 | - | 10 - 11 | 1145 | 105 | |
BC015506.1 | - | 10 - 11 | 1106 | 105 | |
CR542062.1 | - | 10 - 11 | 945 | 105 |