The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003630.2 | NP_003621.1 | 2 - 3 | 466 | 83 |
GenBank mRNA | AB035307.1 | - | 2 - 3 | 350 | 83 |
AJ001625.1 | - | 2 - 3 | 267 | 83 | |
AJ131389.1 | - | 2 - 3 | 263 | 83 | |
AK025644.1 | - | - | 332 | 83 | |
AK300907.1 | - | 2 - 3 | 332 | 83 | |
AY277600.1 | - | 2 - 3 | 223 | 83 | |
BC014551.1 | - | 2 - 3 | 404 | 83 | |
BC015506.1 | - | 2 - 3 | 365 | 83 | |
BP377713.1 | - | 2 - 3 | 466 | 83 | |
CR542062.1 | - | 2 - 3 | 204 | 83 |