The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022078.2 | NP_071361.2 | 6 - 7 | 1389 | 74 |
GenBank mRNA | AK022517.1 | - | 6 - 7 | 1384 | 74 |
AK057930.1 | - | 6 - 7 | 1384 | 74 | |
AK303183.1 | - | 7 - 8 | 1332 | 74 | |
AK308100.1 | - | 6 - 7 | 1252 | 74 | |
AL833886.1 | - | 6 - 7 | 1371 | 74 | |
BC007767.2 | - | 6 - 7 | 1365 | 74 | |
CR936767.1 | - | 6 - 7 | 1389 | 74 |