Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
Chromosome Location
Chr.19: 49690672 - 49713731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136052.2 NP_001129524.1 3 - 4 452 67
NM_001199752.2 NP_001186681.1 3 - 4 516 67
NM_001199753.1 NP_001186682.1 3 - 4 452 67
NM_152359.2 NP_689572.1 2 - 3 383 67
XM_005258505.2 XP_005258562.1 3 - 4 489 67
XM_005258506.4 XP_005258563.1 2 - 3 405 67
XM_006723009.3 XP_006723072.1 2 - 3 294 67
XM_011526438.2 XP_011524740.1 3 - 4 545 67
XM_011526439.2 XP_011524741.1 3 - 4 609 67
XM_011526440.2 XP_011524742.1 3 - 4 461 67
XM_017026265.1 XP_016881754.1 3 - 4 508 67
XM_017026266.1 XP_016881755.1 3 - 4 462 67
XM_017026267.1 XP_016881756.1 3 - 4 451 67
XM_017026268.1 XP_016881757.1 2 - 3 294 67
XM_017026269.1 XP_016881758.1 2 - 3 404 67
XM_017026270.1 XP_016881759.1 2 - 3 172 67
XM_017026271.1 XP_016881760.1 2 - 3 294 67
XM_017026272.1 XP_016881761.1 3 - 4 488 67
XM_017026273.1 XP_016881762.1 3 - 4 473 67
GenBank mRNA AF357970.1 - 2 - 3 341 67
AK290092.1 - 3 - 4 424 67
AK299866.1 - 2 - 3 283 67
AK309087.1 - 3 - 4 444 67
BC029104.1 - 3 - 4 416 67
BP194020.1 - 2 - 3 383 67

View Full Product Details