Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
Chromosome Location
Chr.19: 49690672 - 49713731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136052.2 NP_001129524.1 9 - 10 1167 76
NM_001199752.2 NP_001186681.1 9 - 10 1264 76
NM_001199753.1 NP_001186682.1 9 - 10 1200 76
NM_152359.2 NP_689572.1 8 - 9 1131 76
NR_108072.1 10 - 11 1362 76
XM_005258505.2 XP_005258562.1 9 - 10 1237 76
XM_005258506.4 XP_005258563.1 8 - 9 1153 76
XM_011526438.2 XP_011524740.1 9 - 10 1293 76
XM_011526439.2 XP_011524741.1 9 - 10 1357 76
XM_011526440.2 XP_011524742.1 9 - 10 1209 76
XM_017026265.1 XP_016881754.1 9 - 10 1256 76
XM_017026266.1 XP_016881755.1 9 - 10 1177 76
XM_017026267.1 XP_016881756.1 8 - 9 1027 76
XM_017026268.1 XP_016881757.1 7 - 8 870 76
XM_017026269.1 XP_016881758.1 7 - 8 980 76
XM_017026270.1 XP_016881759.1 7 - 8 748 76
XM_017026271.1 XP_016881760.1 7 - 8 837 76
XM_017026272.1 XP_016881761.1 8 - 9 1064 76
XM_017026273.1 XP_016881762.1 8 - 9 1049 76
GenBank mRNA AB208884.1 - 3 - 4 244 76
AF357970.1 - 8 - 9 1089 76
AK074389.1 - 1 - 2 110 76
AK096516.1 - 10 - 11 1358 76
AK289713.1 - 9 - 10 1256 76
AK290092.1 - 9 - 10 1172 76
AK309087.1 - 8 - 9 1090 76
BC029104.1 - 9 - 10 1131 76

View Full Product Details