Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
Chromosome Location
Chr.19: 49690672 - 49713731 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136052.2 NP_001129524.1 17 - 18 2303 115
NM_001199752.2 NP_001186681.1 17 - 18 2400 115
NM_001199753.1 NP_001186682.1 17 - 18 2336 115
NM_152359.2 NP_689572.1 16 - 17 2267 115
NR_108072.1 18 - 19 2498 115
XM_005258505.2 XP_005258562.1 17 - 18 2373 115
XM_005258506.4 XP_005258563.1 16 - 17 2289 115
XM_006723009.3 XP_006723072.1 14 - 15 2105 115
XM_011526438.2 XP_011524740.1 17 - 18 2429 115
XM_011526439.2 XP_011524741.1 17 - 18 2493 115
XM_011526440.2 XP_011524742.1 16 - 17 2210 115
XM_017026265.1 XP_016881754.1 16 - 17 2257 115
XM_017026266.1 XP_016881755.1 16 - 17 2178 115
XM_017026267.1 XP_016881756.1 16 - 17 2163 115
XM_017026268.1 XP_016881757.1 15 - 16 2006 115
XM_017026269.1 XP_016881758.1 15 - 16 2116 115
XM_017026270.1 XP_016881759.1 15 - 16 1884 115
XM_017026271.1 XP_016881760.1 15 - 16 1973 115
XM_017026272.1 XP_016881761.1 15 - 16 2065 115
XM_017026273.1 XP_016881762.1 15 - 16 2050 115
GenBank mRNA AF357970.1 - 16 - 17 2225 115
AK074389.1 - 9 - 10 1246 115
AK096516.1 - 18 - 19 2494 115
AK289713.1 - 17 - 18 2392 115
AK290092.1 - 17 - 18 2308 115
BC029104.1 - 17 - 18 2267 115

View Full Product Details