The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001870.3 | NP_001861.2 | 5 - 6 | 532 | 155 |
GenBank mRNA | AK313628.1 | - | 5 - 6 | 492 | 155 |
BC012613.1 | - | 5 - 6 | 495 | 155 | |
DR005288.1 | - | 5 - 6 | 482 | 155 | |
M27717.1 | - | 5 - 6 | 480 | 155 | |
S40234.1 | - | 2 - 3 | 153 | 155 |