The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017902.2 | NP_060372.2 | 4 - 5 | 799 | 65 |
XM_011539940.2 | XP_011538242.1 | 4 - 5 | 810 | 65 | |
XM_011539941.1 | XP_011538243.1 | 4 - 5 | 677 | 65 | |
GenBank mRNA | AF395830.1 | - | 4 - 5 | 734 | 65 |
AK000622.1 | - | 4 - 5 | 744 | 65 | |
AK295353.1 | - | 4 - 5 | 826 | 65 | |
AL359615.1 | - | 3 - 4 | 462 | 65 | |
BC007719.2 | - | 4 - 5 | 727 | 65 |