The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015542.3 | NP_056357.1 | 11 - 12 | 2292 | 91 |
NM_080599.2 | NP_542166.1 | 11 - 12 | 2462 | 91 | |
XM_017016034.1 | XP_016871523.1 | 10 - 11 | 1948 | 91 | |
GenBank mRNA | AB037829.1 | - | 10 - 11 | 2661 | 91 |
AF301013.1 | - | 11 - 12 | 2285 | 91 | |
AF318574.1 | - | 11 - 12 | 2262 | 91 | |
AK310925.1 | - | 11 - 12 | 2251 | 91 | |
AL080198.1 | - | 5 - 6 | 462 | 91 | |
AY013249.1 | - | 10 - 11 | 2211 | 91 | |
BC114964.1 | - | 11 - 12 | 2256 | 91 | |
BC115737.1 | - | 11 - 12 | 2256 | 91 |