The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015542.3 | NP_056357.1 | 5 - 6 | 1608 | 141 |
NM_080599.2 | NP_542166.1 | 5 - 6 | 1778 | 141 | |
XM_011519449.2 | XP_011517751.1 | 5 - 6 | 1656 | 141 | |
XM_017016034.1 | XP_016871523.1 | 4 - 5 | 1264 | 141 | |
GenBank mRNA | AB037829.1 | - | 4 - 5 | 1977 | 141 |
AF301013.1 | - | 5 - 6 | 1601 | 141 | |
AF318574.1 | - | 5 - 6 | 1578 | 141 | |
AK096191.1 | - | 6 - 7 | 1518 | 141 | |
AK310925.1 | - | 5 - 6 | 1567 | 141 | |
AY013249.1 | - | 4 - 5 | 1527 | 141 | |
BC114964.1 | - | 5 - 6 | 1572 | 141 | |
BC115737.1 | - | 5 - 6 | 1572 | 141 |