The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282285.1 | NP_001269214.1 | 2 - 3 | 287 | 95 |
NM_001282286.1 | NP_001269215.1 | 2 - 3 | 287 | 95 | |
NM_003845.2 | NP_003836.1 | 10 - 11 | 1183 | 95 | |
GenBank mRNA | AF263541.1 | - | 9 - 10 | 1083 | 95 |
AK292322.1 | - | 10 - 11 | 1177 | 95 | |
AK303744.1 | - | 2 - 3 | 255 | 95 | |
AK308260.1 | - | 12 - 13 | 1404 | 94 | |
AK308702.1 | - | 2 - 3 | 225 | 95 | |
AL561586.3 | - | 2 - 3 | 287 | 95 | |
BC031244.1 | - | 10 - 11 | 1177 | 95 | |
GU139741.1 | - | 2 - 3 | 219 | 95 | |
GU139742.1 | - | 2 - 3 | 200 | 95 | |
GU139743.1 | - | 9 - 10 | 1167 | 95 | |
GU139745.1 | - | - | 853 | 95 | |
Y09305.1 | - | 2 - 3 | 181 | 95 |