The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243374.1 | NP_001230303.1 | 7 - 8 | 1240 | 99 |
NM_001829.3 | NP_001820.2 | 8 - 9 | 1572 | 99 | |
NM_173872.3 | NP_776297.2 | 8 - 9 | 1572 | 99 | |
XM_005262726.2 | XP_005262783.1 | 7 - 8 | 1112 | 99 | |
XM_011531586.2 | XP_011529888.1 | 7 - 8 | 1622 | 99 | |
GenBank mRNA | AB019542.1 | - | 8 - 9 | 1100 | 99 |
AF029346.1 | - | 7 - 8 | 1013 | 99 | |
AF172729.1 | - | 7 - 8 | 1013 | 99 | |
AK127483.1 | - | 7 - 8 | 1240 | 99 | |
BC136510.1 | - | 9 - 10 | 1347 | 99 | |
BX647119.1 | - | 8 - 9 | 1522 | 99 | |
X78520.1 | - | 8 - 9 | 1501 | 99 |