The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243372.1 | NP_001230301.1 | 8 - 9 | 2040 | 110 |
NM_001243374.1 | NP_001230303.1 | 8 - 9 | 1789 | 110 | |
NM_001829.3 | NP_001820.2 | 9 - 10 | 2121 | 110 | |
NM_173872.3 | NP_776297.2 | 9 - 10 | 2121 | 110 | |
XM_005262726.2 | XP_005262783.1 | 8 - 9 | 1661 | 110 | |
XM_011531586.2 | XP_011529888.1 | 8 - 9 | 2171 | 110 | |
GenBank mRNA | AB019542.1 | - | 9 - 10 | 1649 | 110 |
AF029346.1 | - | 8 - 9 | 1562 | 110 | |
AF172729.1 | - | 8 - 9 | 1562 | 110 | |
AK127483.1 | - | 8 - 9 | 1789 | 110 | |
AK304362.1 | - | 8 - 9 | 1974 | 110 | |
BC136510.1 | - | 10 - 11 | 1896 | 110 | |
BX647119.1 | - | 9 - 10 | 2071 | 110 | |
X78520.1 | - | 9 - 10 | 2050 | 110 |