The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127218.2 | NP_001120690.1 | 8 - 9 | 1303 | 79 |
NM_001256879.1 | NP_001243808.1 | 9 - 10 | 1664 | 79 | |
NM_006230.3 | NP_006221.2 | 8 - 9 | 1130 | 79 | |
XM_011515431.1 | XP_011513733.1 | 8 - 9 | 1126 | 79 | |
GenBank mRNA | AK292347.1 | - | 9 - 10 | 1664 | 79 |
AK298168.1 | - | 8 - 9 | 994 | 79 | |
AK312294.1 | - | 8 - 9 | 1263 | 79 | |
BC000459.2 | - | 8 - 9 | 1093 | 79 | |
BC030001.2 | - | 7 - 8 | 1070 | 79 | |
U21090.1 | - | 8 - 9 | 1092 | 79 |