The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144888.1 | NP_001138360.1 | 6 - 7 | 599 | 78 |
NM_006340.2 | NP_006331.1 | 6 - 7 | 599 | 78 | |
NM_017450.2 | NP_059344.1 | 6 - 7 | 599 | 78 | |
NM_017451.2 | NP_059345.1 | 6 - 7 | 599 | 78 | |
XM_005256943.1 | XP_005257000.1 | 7 - 8 | 698 | 78 | |
XM_005256944.1 | XP_005257001.1 | 7 - 8 | 698 | 78 | |
XM_005256945.1 | XP_005257002.1 | 7 - 8 | 698 | 78 | |
XM_005256948.3 | XP_005257005.1 | 6 - 7 | 599 | 78 | |
XM_006721635.1 | XP_006721698.1 | 7 - 8 | 698 | 78 | |
XM_006721636.1 | XP_006721699.1 | 7 - 8 | 698 | 78 | |
XM_006721637.2 | XP_006721700.1 | 6 - 7 | 599 | 78 | |
XM_011524193.1 | XP_011522495.1 | 7 - 8 | 698 | 78 | |
XM_011524194.1 | XP_011522496.1 | 7 - 8 | 698 | 78 | |
XM_017024017.1 | XP_016879506.1 | 5 - 6 | 1356 | 78 | |
XM_017024018.1 | XP_016879507.1 | 4 - 5 | 818 | 78 | |
GenBank mRNA | AB015019.1 | - | 6 - 7 | 584 | 78 |
AB015020.1 | - | 6 - 7 | 584 | 78 | |
AB017119.1 | - | 6 - 7 | 584 | 78 | |
AB017120.1 | - | 6 - 7 | 584 | 78 | |
AK023688.1 | - | 7 - 8 | 684 | 78 | |
AK056888.1 | - | 2 - 3 | 771 | 78 | |
AK222670.1 | - | 6 - 7 | 595 | 78 | |
AK225612.1 | - | 6 - 7 | 596 | 78 | |
AK297644.1 | - | 6 - 7 | 528 | 78 | |
AK298629.1 | - | 4 - 5 | 434 | 78 | |
BC002495.2 | - | 6 - 7 | 574 | 78 | |
BC014020.2 | - | 6 - 7 | 586 | 78 | |
BC032559.1 | - | 6 - 7 | 597 | 78 | |
U70669.1 | - | 6 - 7 | 584 | 78 |