The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002741.3 | NP_002732.3 | 1 - 2 | 190 | 74 |
GenBank mRNA | BC094766.1 | - | 1 - 2 | 221 | 74 |
S75546.1 | - | 1 - 2 | 106 | 74 | |
U33053.1 | - | 1 - 2 | 106 | 74 |