Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKN1
Entrez Gene ID
5585
Gene Name
protein kinase N1
Gene Aliases
DBK, PAK-1, PAK1, PKN, PKN-ALPHA, PRK1, PRKCL1
Chromosome Location
Chr.19: 14433354 - 14471867 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002741.3 NP_002732.3 2 - 3 491 98
NM_213560.1 NP_998725.1 2 - 3 358 98
XM_011528128.1 XP_011526430.1 2 - 3 374 98
GenBank mRNA AK123007.1 - 2 - 3 372 98
AK292130.1 - 2 - 3 355 98
AK313886.1 - 2 - 3 331 98
BC040061.1 - 2 - 3 488 98
BC094766.1 - 2 - 3 522 98
D26181.1 - 2 - 3 359 98
S75546.1 - 2 - 3 407 98
U33053.1 - 2 - 3 407 98

View Full Product Details