The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300823.1 | NP_001287752.1 | 14 - 15 | 1890 | 90 |
NM_001300824.1 | NP_001287753.1 | 13 - 14 | 1804 | 90 | |
NM_018699.3 | NP_061169.2 | 15 - 16 | 1983 | 90 | |
XM_011531562.2 | XP_011529864.1 | 15 - 16 | 1999 | 90 | |
XM_011531563.2 | XP_011529865.1 | 14 - 15 | 1906 | 90 | |
XM_011531567.2 | XP_011529869.1 | 14 - 15 | 1913 | 90 | |
XM_011531570.2 | XP_011529872.1 | 13 - 14 | 1616 | 90 | |
XM_017007668.1 | XP_016863157.1 | 15 - 16 | 2492 | 90 | |
XM_017007669.1 | XP_016863158.1 | 14 - 15 | 1880 | 90 | |
XM_017007671.1 | XP_016863160.1 | 9 - 10 | 1090 | 90 | |
GenBank mRNA | AF272897.1 | - | 15 - 16 | 1821 | 90 |
AK314293.1 | - | 15 - 16 | 2229 | 90 | |
BC121037.1 | - | 14 - 15 | 1677 | 90 | |
BC121038.1 | - | 13 - 14 | 1591 | 90 |