The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002737.2 | NP_002728.1 | 12 - 13 | 1424 | 93 |
XM_017024836.1 | XP_016880325.1 | 12 - 13 | 1592 | 93 | |
XM_017024837.1 | XP_016880326.1 | 11 - 12 | 1255 | 93 | |
XM_017024838.1 | XP_016880327.1 | 12 - 13 | 1351 | 93 | |
XM_017024839.1 | XP_016880328.1 | 11 - 12 | 2290 | 93 | |
XM_017024840.1 | XP_016880329.1 | 12 - 13 | 1333 | 93 | |
GenBank mRNA | AB209475.1 | - | 13 - 14 | 1433 | 93 |
AB451258.1 | - | 12 - 13 | 1380 | 93 | |
AB451383.1 | - | 12 - 13 | 1380 | 93 | |
AY633609.1 | - | 12 - 13 | 1385 | 93 | |
BC109273.1 | - | 12 - 13 | 1406 | 93 | |
BC109274.1 | - | 12 - 13 | 1406 | 93 | |
LC151409.1 | - | 13 - 14 | 1380 | 93 | |
X52479.1 | - | 12 - 13 | 1407 | 93 |