Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD3
Entrez Gene ID
51133
Gene Name
potassium channel tetramerization domain containing 3
Gene Aliases
NY-REN-45
Chromosome Location
Chr.1: 215567185 - 215621821 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319294.1 NP_001306223.1 13 - 14 1615 127
NM_001319295.1 NP_001306224.1 13 - 14 1687 127
NM_016121.4 NP_057205.2 13 - 14 1615 127
XM_005273158.1 XP_005273215.1 13 - 14 1881 127
GenBank mRNA AK055925.1 - 7 - 8 892 127
AK316272.1 - 9 - 10 1016 127
AL834389.1 - 7 - 8 839 127
BC013868.2 - 13 - 14 1602 127
BC094879.1 - 13 - 14 1564 127
BC117188.1 - 13 - 14 1497 127
BC126173.1 - 13 - 14 1497 127

View Full Product Details