The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304803.1 | NP_001291732.1 | 5 - 6 | 499 | 72 |
NM_021947.2 | NP_068766.1 | 6 - 7 | 671 | 72 | |
XM_006721565.3 | XP_006721628.1 | 6 - 7 | 703 | 72 | |
XM_006721566.3 | XP_006721629.1 | 6 - 7 | 1881 | 72 | |
XM_011523974.2 | XP_011522276.1 | 6 - 7 | 810 | 72 | |
GenBank mRNA | AF169974.1 | - | 5 - 6 | 608 | 72 |
AK023169.1 | - | 6 - 7 | 660 | 72 | |
AK223120.1 | - | 6 - 7 | 664 | 72 | |
AL834378.1 | - | 4 - 5 | 436 | 72 | |
AY034081.1 | - | 5 - 6 | 592 | 72 | |
BC074728.2 | - | 5 - 6 | 610 | 72 | |
CR457300.1 | - | 5 - 6 | 592 | 72 | |
DA297969.1 | - | 5 - 6 | 488 | 72 |