Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGPS
Entrez Gene ID
8540
Gene Name
alkylglycerone phosphate synthase
Gene Aliases
ADAP-S, ADAS, ADHAPS, ADPS, ALDHPSY, RCDP3
Chromosome Location
Chr.2: 177392743 - 177543836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003659.3 NP_003650.1 20 - 20 7310 122
XM_011512041.2 XP_011510343.1 20 - 20 7092 122
XM_011512042.1 XP_011510344.1 20 - 20 7259 122
XM_011512043.2 XP_011510345.1 16 - 16 7896 122

View Full Product Details