Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC16A
Entrez Gene ID
9919
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases
KIAA0310, SEC16L, p250
Chromosome Location
Chr.9: 136440096 - 136484552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276418.1 NP_001263347.1 11 - 12 5211 62
NM_014866.1 NP_055681.1 12 - 13 5201 62
XM_005266134.2 XP_005266191.1 11 - 12 5064 62
XM_011519246.2 XP_011517548.1 12 - 13 5210 62
XM_011519247.1 XP_011517549.1 11 - 12 5067 62
XM_011519248.2 XP_011517550.1 11 - 12 5041 62
XM_011519249.1 XP_011517551.1 11 - 12 5064 62
XM_011519250.2 XP_011517552.1 11 - 12 5064 62
XM_011519251.1 XP_011517553.1 11 - 12 5064 62
XM_011519252.2 XP_011517554.1 11 - 12 5064 62
XM_011519253.1 XP_011517555.1 11 - 12 5064 62
XM_011519254.1 XP_011517556.1 11 - 12 5064 62
XM_011519255.2 XP_011517557.1 11 - 12 5064 62
XM_011519256.1 XP_011517558.1 11 - 12 5064 62
XM_011519257.1 XP_011517559.1 11 - 12 5064 62
XM_011519258.1 XP_011517560.1 11 - 12 5064 62
XM_011519259.1 XP_011517561.1 11 - 12 5064 62
XM_011519260.2 XP_011517562.1 11 - 12 5048 62
XM_011519262.1 XP_011517564.1 12 - 13 4353 62
XM_011519263.2 XP_011517565.1 12 - 13 4353 62
XM_011519264.2 XP_011517566.1 11 - 12 5064 62
XM_017015363.1 XP_016870852.1 11 - 12 5064 62
GenBank mRNA AB002308.4 - 12 - 13 5201 62
AB593115.1 - 11 - 12 5211 62
BC008332.1 - 1 - 2 74 62
BC028183.1 - 7 - 8 1068 62
BC125018.1 - 10 - 11 3773 62
BC125019.1 - 11 - 12 3062 62
DQ903855.1 - 10 - 11 4358 62

View Full Product Details