Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC16A
Entrez Gene ID
9919
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases
KIAA0310, SEC16L, p250
Chromosome Location
Chr.9: 136440096 - 136484552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276418.1 NP_001263347.1 16 - 17 5729 97
NM_014866.1 NP_055681.1 17 - 18 5719 97
XM_005266134.2 XP_005266191.1 16 - 17 5582 97
XM_011519246.2 XP_011517548.1 17 - 18 5728 97
XM_011519247.1 XP_011517549.1 16 - 17 5585 97
XM_011519248.2 XP_011517550.1 16 - 17 5559 97
XM_011519249.1 XP_011517551.1 16 - 17 5582 97
XM_011519250.2 XP_011517552.1 16 - 17 5582 97
XM_011519251.1 XP_011517553.1 16 - 17 5582 97
XM_011519252.2 XP_011517554.1 16 - 17 5582 97
XM_011519253.1 XP_011517555.1 16 - 17 5582 97
XM_011519254.1 XP_011517556.1 16 - 17 5582 97
XM_011519255.2 XP_011517557.1 16 - 17 5582 97
XM_011519256.1 XP_011517558.1 16 - 17 5582 97
XM_011519257.1 XP_011517559.1 16 - 17 5582 97
XM_011519259.1 XP_011517561.1 16 - 17 5582 97
XM_011519260.2 XP_011517562.1 16 - 17 5566 97
XM_011519261.1 XP_011517563.1 13 - 14 5135 97
XM_011519262.1 XP_011517564.1 17 - 18 4871 97
XM_011519263.2 XP_011517565.1 17 - 18 4871 97
XM_011519264.2 XP_011517566.1 16 - 17 5582 97
XM_017015363.1 XP_016870852.1 16 - 17 5582 97
GenBank mRNA AB002308.4 - 17 - 18 5719 97
AB593115.1 - 16 - 17 5729 97
BC008332.1 - 6 - 7 592 97
BC028183.1 - 12 - 13 1586 97
BC125018.1 - 15 - 16 4291 97
BC125019.1 - 16 - 17 3580 97
DQ903855.1 - 15 - 16 4876 97

View Full Product Details