The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001276418.1 | NP_001263347.1 | 28 - 29 | 7254 | 79 |
NM_014866.1 | NP_055681.1 | 31 - 32 | 7313 | 79 | |
XM_005266134.2 | XP_005266191.1 | 30 - 31 | 7242 | 79 | |
XM_011519246.2 | XP_011517548.1 | 31 - 32 | 7388 | 79 | |
XM_011519247.1 | XP_011517549.1 | 30 - 31 | 7245 | 79 | |
XM_011519248.2 | XP_011517550.1 | 30 - 31 | 7219 | 79 | |
XM_011519249.1 | XP_011517551.1 | 30 - 31 | 7233 | 79 | |
XM_011519251.1 | XP_011517553.1 | 29 - 30 | 7182 | 79 | |
XM_011519252.2 | XP_011517554.1 | 30 - 31 | 7176 | 79 | |
XM_011519253.1 | XP_011517555.1 | 29 - 30 | 7173 | 79 | |
XM_011519254.1 | XP_011517556.1 | 29 - 30 | 7167 | 79 | |
XM_011519255.2 | XP_011517557.1 | 29 - 30 | 7116 | 79 | |
XM_011519256.1 | XP_011517558.1 | 28 - 29 | 7107 | 79 | |
XM_011519257.1 | XP_011517559.1 | 29 - 30 | 7101 | 79 | |
XM_011519258.1 | XP_011517560.1 | 29 - 30 | 7101 | 79 | |
XM_011519259.1 | XP_011517561.1 | 28 - 29 | 7041 | 79 | |
XM_011519260.2 | XP_011517562.1 | 30 - 31 | 7226 | 79 | |
XM_011519261.1 | XP_011517563.1 | 27 - 28 | 6795 | 79 | |
XM_011519262.1 | XP_011517564.1 | 31 - 32 | 6531 | 79 | |
XM_011519263.2 | XP_011517565.1 | 30 - 31 | 6405 | 79 | |
XM_017015363.1 | XP_016870852.1 | 29 - 30 | 7158 | 79 | |
GenBank mRNA | AB002308.4 | - | 31 - 32 | 7313 | 79 |
AB593115.1 | - | 28 - 29 | 7254 | 79 | |
AK074565.1 | - | 5 - 6 | 544 | 79 | |
AK094756.1 | - | 9 - 10 | 1351 | 79 | |
AK124843.1 | - | 2 - 3 | 666 | 79 | |
AK301371.1 | - | 7 - 8 | 583 | 79 | |
AK302210.1 | - | 6 - 7 | 576 | 79 | |
AK316194.1 | - | 5 - 6 | 478 | 79 | |
BC001404.2 | - | 14 - 15 | 1449 | 79 | |
BC008332.1 | - | 20 - 21 | 2186 | 79 | |
BC016873.1 | - | 5 - 6 | 406 | 79 | |
BC028183.1 | - | 25 - 26 | 3120 | 79 | |
BC098454.1 | - | 14 - 15 | 1542 | 79 | |
BC125018.1 | - | 27 - 28 | 5816 | 79 | |
BC125019.1 | - | 29 - 30 | 5114 | 79 | |
DQ903855.1 | - | 28 - 29 | 6395 | 79 |