The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256045.1 | NP_001242974.1 | 4 - 5 | 915 | 71 |
NM_006219.2 | NP_006210.1 | 13 - 14 | 2049 | 71 | |
XM_005247530.2 | XP_005247587.1 | 15 - 16 | 2398 | 71 | |
XM_006713659.3 | XP_006713722.1 | 14 - 15 | 2269 | 71 | |
XM_011512895.2 | XP_011511197.1 | 16 - 17 | 2394 | 71 | |
XM_011512896.1 | XP_011511198.1 | 11 - 12 | 1663 | 71 | |
XM_017006619.1 | XP_016862108.1 | 14 - 15 | 2072 | 71 | |
XM_017006620.1 | XP_016862109.1 | 13 - 14 | 1819 | 71 | |
XM_017006621.1 | XP_016862110.1 | 12 - 13 | 1768 | 71 | |
GenBank mRNA | AK293754.1 | - | 4 - 5 | 915 | 71 |
AK302937.1 | - | 6 - 7 | 928 | 71 | |
BC114432.1 | - | 13 - 14 | 2037 | 71 | |
CR749357.1 | - | 6 - 7 | 619 | 71 | |
S67334.1 | - | 13 - 14 | 2033 | 71 |