The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256045.1 | NP_001242974.1 | 12 - 13 | 1957 | 82 |
NM_006219.2 | NP_006210.1 | 21 - 22 | 3091 | 82 | |
XM_005247530.2 | XP_005247587.1 | 23 - 24 | 3440 | 82 | |
XM_006713659.3 | XP_006713722.1 | 22 - 23 | 3311 | 82 | |
XM_011512895.2 | XP_011511197.1 | 24 - 25 | 3436 | 82 | |
XM_011512896.1 | XP_011511198.1 | 19 - 20 | 2705 | 82 | |
XM_017006619.1 | XP_016862108.1 | 22 - 23 | 3114 | 82 | |
XM_017006620.1 | XP_016862109.1 | 21 - 22 | 2861 | 82 | |
XM_017006621.1 | XP_016862110.1 | 20 - 21 | 2810 | 82 | |
GenBank mRNA | AK293754.1 | - | 12 - 13 | 1957 | 82 |
AK302937.1 | - | 14 - 15 | 1970 | 82 | |
BC003393.1 | - | 6 - 7 | 724 | 82 | |
BC022049.2 | - | 2 - 3 | 282 | 82 | |
BC114432.1 | - | 21 - 22 | 3079 | 82 | |
CR749357.1 | - | 14 - 15 | 1676 | 82 | |
S67334.1 | - | 21 - 22 | 3075 | 82 |