The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083615.3 | NP_001077084.2 | 13 - 14 | 1520 | 97 |
NM_138995.4 | NP_620482.3 | 13 - 14 | 1520 | 97 | |
NR_045682.1 | 13 - 14 | 1520 | 97 | ||
NR_045683.1 | 13 - 14 | 1520 | 97 | ||
NR_045684.1 | 13 - 14 | 1520 | 97 | ||
XM_006712299.3 | XP_006712362.1 | 13 - 14 | 1408 | 97 | |
XM_011510654.2 | XP_011508956.1 | 13 - 14 | 1408 | 97 | |
XM_011510655.2 | XP_011508957.1 | 13 - 14 | 1408 | 97 | |
XM_011510656.2 | XP_011508958.1 | 7 - 8 | 783 | 97 | |
XM_011510657.2 | XP_011508959.1 | 6 - 7 | 655 | 97 | |
XM_011510658.2 | XP_011508960.1 | 7 - 8 | 964 | 97 | |
GenBank mRNA | AF369908.1 | - | 13 - 14 | 1520 | 97 |
AF391554.1 | - | 13 - 14 | 1520 | 97 | |
AF391555.1 | - | 13 - 14 | 1520 | 97 | |
AF391556.1 | - | 13 - 14 | 1520 | 97 | |
AF391557.1 | - | 13 - 14 | 1520 | 97 | |
AF391558.1 | - | 13 - 14 | 1520 | 97 | |
AK055778.1 | - | 13 - 14 | 1408 | 97 | |
BC136620.1 | - | 13 - 14 | 1457 | 97 | |
BC144307.1 | - | 13 - 14 | 1457 | 97 | |
BX647895.1 | - | 11 - 12 | 1245 | 97 |