The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083615.3 | NP_001077084.2 | 22 - 23 | 2791 | 76 |
NM_138995.4 | NP_620482.3 | 22 - 23 | 2791 | 76 | |
NR_045682.1 | 22 - 23 | 2791 | 76 | ||
NR_045683.1 | 22 - 23 | 2791 | 76 | ||
NR_045684.1 | 22 - 23 | 2791 | 76 | ||
XM_006712299.3 | XP_006712362.1 | 22 - 23 | 2679 | 76 | |
XM_011510654.2 | XP_011508956.1 | 22 - 23 | 2679 | 76 | |
XM_011510655.2 | XP_011508957.1 | 22 - 23 | 2679 | 76 | |
XM_011510656.2 | XP_011508958.1 | 16 - 17 | 2054 | 76 | |
XM_011510657.2 | XP_011508959.1 | 15 - 16 | 1926 | 76 | |
XM_011510658.2 | XP_011508960.1 | 16 - 17 | 2235 | 76 | |
GenBank mRNA | AF369908.1 | - | 22 - 23 | 2791 | 76 |
AF391554.1 | - | 22 - 23 | 2791 | 76 | |
AF391555.1 | - | 22 - 23 | 2791 | 76 | |
AF391556.1 | - | 22 - 23 | 2791 | 76 | |
AF391557.1 | - | 22 - 23 | 2791 | 76 | |
AF391558.1 | - | 22 - 23 | 2791 | 76 | |
AK055778.1 | - | 22 - 23 | 2679 | 76 | |
BC136620.1 | - | 22 - 23 | 2728 | 76 | |
BC144307.1 | - | 22 - 23 | 2728 | 76 | |
BX647895.1 | - | 20 - 21 | 2516 | 76 |