The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083615.3 | NP_001077084.2 | 33 - 34 | 3945 | 138 |
NM_138995.4 | NP_620482.3 | 34 - 35 | 4026 | 138 | |
NR_045682.1 | 35 - 36 | 4148 | 138 | ||
NR_045683.1 | 35 - 36 | 4057 | 138 | ||
NR_045684.1 | 34 - 35 | 3976 | 138 | ||
XM_006712299.3 | XP_006712362.1 | 34 - 35 | 3914 | 138 | |
XM_011510654.2 | XP_011508956.1 | 35 - 36 | 4022 | 138 | |
XM_011510655.2 | XP_011508957.1 | 33 - 34 | 3833 | 138 | |
XM_011510656.2 | XP_011508958.1 | 29 - 30 | 3397 | 138 | |
XM_011510657.2 | XP_011508959.1 | 28 - 29 | 3269 | 138 | |
XM_011510658.2 | XP_011508960.1 | 29 - 30 | 3578 | 138 | |
GenBank mRNA | AF369908.1 | - | 35 - 36 | 4057 | 138 |
AF391554.1 | - | 34 - 35 | 3976 | 138 | |
AF391555.1 | - | 34 - 35 | 4026 | 138 | |
AF391556.1 | - | 33 - 34 | 3945 | 138 | |
BC136620.1 | - | 34 - 35 | 3963 | 138 | |
BC144307.1 | - | 34 - 35 | 3963 | 138 | |
BX647895.1 | - | 27 - 28 | 3111 | 138 |