The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018699.3 | NP_061169.2 | 6 - 7 | 998 | 94 |
XM_005262708.3 | XP_005262765.1 | 6 - 7 | 981 | 94 | |
XM_011531562.2 | XP_011529864.1 | 6 - 7 | 981 | 94 | |
XM_011531564.2 | XP_011529866.1 | 6 - 7 | 981 | 94 | |
XM_011531565.2 | XP_011529867.1 | 6 - 7 | 981 | 94 | |
XM_011531566.2 | XP_011529868.1 | 6 - 7 | 981 | 94 | |
XM_011531567.2 | XP_011529869.1 | 6 - 7 | 981 | 94 | |
XM_011531568.2 | XP_011529870.1 | 6 - 7 | 981 | 94 | |
XM_011531569.2 | XP_011529871.1 | 6 - 7 | 981 | 94 | |
XM_017007668.1 | XP_016863157.1 | 6 - 7 | 1474 | 94 | |
XM_017007669.1 | XP_016863158.1 | 6 - 7 | 981 | 94 | |
XM_017007670.1 | XP_016863159.1 | 6 - 7 | 981 | 94 | |
GenBank mRNA | BM563688.1 | - | 2 - 3 | 283 | 94 |