The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002613.4 | NP_002604.1 | 13 - 14 | 1703 | 105 |
NM_031268.5 | NP_112558.2 | 10 - 11 | 1322 | 105 | |
XM_011522521.2 | XP_011520823.1 | 13 - 14 | 5391 | 105 | |
XM_011522523.2 | XP_011520825.1 | 10 - 11 | 5015 | 105 | |
GenBank mRNA | AF017995.1 | - | 13 - 14 | 1634 | 105 |
AK131060.1 | - | 13 - 14 | 1637 | 105 | |
AK222581.1 | - | 13 - 14 | 1646 | 105 | |
BC006339.2 | - | 10 - 11 | 1283 | 105 | |
BC012103.1 | - | 13 - 14 | 1663 | 105 | |
BC033494.1 | - | 10 - 11 | 1235 | 105 | |
CR536517.1 | - | 13 - 14 | 1554 | 105 | |
Y15056.1 | - | 13 - 14 | 1633 | 105 |