Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC16
Entrez Gene ID
57544
Gene Name
thioredoxin domain containing 16
Gene Aliases
ERp90, KIAA1344
Chromosome Location
Chr.14: 52430590 - 52552583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160047.1 NP_001153519.1 18 - 19 2277 134
NM_020784.2 NP_065835.2 18 - 19 2292 134
GenBank mRNA AB037765.2 - 18 - 19 2204 134
AK022563.1 - 4 - 5 478 134
AK097748.1 - 19 - 20 2181 134
BC137081.1 - 18 - 19 2172 134
BC137082.1 - 18 - 19 2172 134
BC142650.1 - 18 - 19 2215 134
BC144469.1 - 18 - 19 2157 134
BC150205.1 - 18 - 19 2213 134
BC152428.1 - 18 - 19 2204 134

View Full Product Details