The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199342.1 | NP_001186271.1 | 1 - 2 | 204 | 67 |
GenBank mRNA | BQ877039.1 | - | 1 - 2 | 145 | 67 |