Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHDC1
Entrez Gene ID
122773
Gene Name
kelch domain containing 1
Gene Aliases
MST025
Chromosome Location
Chr.14: 49693105 - 49753152 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172193.2 NP_751943.1 11 - 12 1067 91
GenBank mRNA AF111806.1 - 11 - 12 1005 91
AK127202.1 - 11 - 12 990 91
AL833437.1 - 6 - 7 497 91
BC101595.1 - 11 - 12 1067 91
BC101597.1 - 11 - 12 1067 91

View Full Product Details