The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160408.1 | NP_001153880.1 | 9 - 10 | 1104 | 149 |
NM_003324.4 | NP_003315.2 | 9 - 10 | 1104 | 149 | |
GenBank mRNA | AF045583.1 | - | 9 - 10 | 1068 | 149 |
AK024246.1 | - | 9 - 10 | 1104 | 149 | |
AK307434.1 | - | 7 - 8 | 807 | 149 | |
BC032587.1 | - | 9 - 10 | 1077 | 149 |