Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SVIL
- Entrez Gene ID
-
6840
- Gene Name
- supervillin
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 29457338 - 29737001 on Build GRCh38
View Full Product Details