The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323599.1 | NP_001310528.1 | 2 - 3 | 422 | 141 |
NM_001323600.1 | NP_001310529.1 | 2 - 3 | 275 | 141 | |
NM_003174.3 | NP_003165.2 | 2 - 3 | 156 | 141 | |
XM_005252570.2 | XP_005252627.1 | 2 - 3 | 356 | 141 | |
XM_005252571.3 | XP_005252628.1 | 2 - 3 | 282 | 141 | |
XM_011519634.1 | XP_011517936.1 | 2 - 3 | 414 | 141 | |
GenBank mRNA | AF051850.1 | - | 2 - 3 | 156 | 141 |
AK307640.1 | - | 2 - 3 | 356 | 141 |