The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199340.1 | NP_001186269.1 | 2 - 3 | 209 | 62 |
NM_001199342.1 | NP_001186271.1 | 2 - 3 | 308 | 62 | |
NM_001199343.1 | NP_001186272.1 | 2 - 3 | 291 | 62 | |
GenBank mRNA | BG758380.1 | - | 2 - 3 | 227 | 62 |
BG827384.1 | - | 1 - 2 | 111 | 62 | |
BQ877039.1 | - | 2 - 3 | 249 | 62 |