Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL9A1
Entrez Gene ID
1297
Gene Name
collagen type IX alpha 1
Gene Aliases
DJ149L1.1.2, EDM6, MED, STL4
Chromosome Location
Chr.6: 70216040 - 70303083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001851.4 NP_001842.3 31 - 32 2199 64
NM_078485.3 NP_511040.2 25 - 26 1467 64
XM_011535429.2 XP_011533731.1 32 - 33 2229 64
XM_011535430.2 XP_011533732.1 26 - 27 1495 64
XM_017010246.1 XP_016865735.1 29 - 30 1586 64
XM_017010247.1 XP_016865736.1 23 - 24 1442 64
GenBank mRNA AK097582.1 - 3 - 4 745 64
AK125738.1 - 20 - 21 1060 64
BC008620.1 - 25 - 26 1394 64
BC063646.1 - 25 - 26 1441 64
X54412.1 - 31 - 32 2171 64

View Full Product Details