Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL9A1
Entrez Gene ID
1297
Gene Name
collagen type IX alpha 1
Gene Aliases
DJ149L1.1.2, EDM6, MED, STL4
Chromosome Location
Chr.6: 70216040 - 70303083 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001851.4 NP_001842.3 35 - 36 2478 86
NM_078485.3 NP_511040.2 29 - 30 1746 86
XM_011535429.2 XP_011533731.1 36 - 37 2508 86
XM_011535430.2 XP_011533732.1 30 - 31 1774 86
XM_017010246.1 XP_016865735.1 33 - 34 1865 86
XM_017010247.1 XP_016865736.1 27 - 28 1721 86
GenBank mRNA AK097582.1 - 7 - 8 1024 86
AK125738.1 - 24 - 25 1339 86
BC063646.1 - 29 - 30 1720 86
X54412.1 - 35 - 36 2450 86

View Full Product Details