The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000246.3 | NP_000237.2 | 17 - 18 | 3367 | 64 |
NM_001286402.1 | NP_001273331.1 | 17 - 18 | 3370 | 64 | |
NM_001286403.1 | NP_001273332.1 | 15 - 16 | 1615 | 64 | |
NR_104444.1 | 15 - 16 | 1557 | 64 | ||
XM_006720880.3 | XP_006720943.2 | 17 - 18 | 3548 | 64 | |
XM_011522484.2 | XP_011520786.1 | 17 - 18 | 3548 | 64 | |
XM_011522485.2 | XP_011520787.1 | 17 - 18 | 3548 | 64 | |
XM_011522487.2 | XP_011520789.1 | 16 - 17 | 3285 | 64 | |
XM_011522488.2 | XP_011520790.1 | 17 - 18 | 4779 | 64 | |
XM_011522489.2 | XP_011520791.1 | 16 - 17 | 3282 | 64 | |
XM_011522490.2 | XP_011520792.1 | 17 - 18 | 4777 | 64 | |
XM_011522492.2 | XP_011520794.1 | 17 - 18 | 3496 | 64 | |
XM_011522493.2 | XP_011520795.1 | 17 - 18 | 3493 | 64 | |
XM_011522494.2 | XP_011520796.1 | 17 - 18 | 3260 | 64 | |
XM_011522495.2 | XP_011520797.1 | 16 - 17 | 3351 | 64 | |
XM_011522496.2 | XP_011520798.1 | 16 - 17 | 3348 | 64 | |
GenBank mRNA | AF410154.1 | - | 17 - 18 | 4248 | 64 |
AY084054.1 | - | 15 - 16 | 1605 | 64 | |
AY699071.1 | - | 16 - 17 | 3165 | 64 | |
U18259.1 | - | 17 - 18 | 3372 | 64 | |
X74301.1 | - | 17 - 18 | 3349 | 64 |