The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002553.3 | NP_002544.1 | 1 - 2 | 210 | 149 |
NM_181747.3 | NP_859531.1 | 1 - 2 | 210 | 149 | |
XM_011516273.2 | XP_011514575.1 | 1 - 2 | 204 | 149 | |
GenBank mRNA | AF047599.1 | - | 1 - 2 | 145 | 149 |
AF049127.1 | - | 1 - 2 | 141 | 149 | |
AF081459.1 | - | 1 - 2 | 120 | 149 | |
AK223361.1 | - | 1 - 2 | 210 | 149 | |
AK292285.1 | - | 1 - 2 | 186 | 149 | |
AK313596.1 | - | 1 - 2 | 167 | 149 | |
BC023652.2 | - | 1 - 2 | 148 | 149 | |
U92538.1 | - | 1 - 2 | 155 | 149 |