The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014840.2 | NP_055655.1 | 2 - 3 | 1738 | 91 |
GenBank mRNA | AB011109.1 | - | 2 - 3 | 1738 | 91 |
AK027302.1 | - | 3 - 4 | 599 | 91 | |
AK289992.1 | - | 2 - 3 | 418 | 91 | |
BC152462.1 | - | 2 - 3 | 1738 | 91 |