Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WAC
Entrez Gene ID
51322
Gene Name
WW domain containing adaptor with coiled-coil
Gene Aliases
BM-016, DESSH, PRO1741, Wwp4
Chromosome Location
Chr.10: 28532426 - 28623112 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016628.4 NP_057712.2 11 - 12 1642 60
NM_100264.2 NP_567822.1 11 - 12 1933 60
NM_100486.3 NP_567823.1 10 - 11 1333 60
XM_011519491.2 XP_011517793.1 11 - 12 2243 60
XM_017016317.1 XP_016871806.1 10 - 11 1444 60
GenBank mRNA AF208858.1 - 4 - 5 567 60
AK055852.1 - 11 - 12 1750 60
AK223600.1 - 11 - 12 1640 60
AK225846.1 - 11 - 12 1746 60
AK290174.1 - 11 - 12 1647 60
BC004258.1 - 11 - 12 1633 60
BC010356.1 - 6 - 7 784 60

View Full Product Details