Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ORC5
Entrez Gene ID
5001
Gene Name
origin recognition complex subunit 5
Gene Aliases
ORC5L, ORC5P, ORC5T, PPP1R117
Chromosome Location
Chr.7: 104126341 - 104208047 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002553.3 NP_002544.1 13 - 14 1404 108
GenBank mRNA AF047599.1 - 13 - 14 1339 108
AF049127.1 - 13 - 14 1335 108
AK223361.1 - 13 - 14 1404 108
AK292285.1 - 13 - 14 1380 108
AK302122.1 - 14 - 15 1491 108
BC023652.2 - 13 - 14 1342 108
U92538.1 - 13 - 14 1349 108

View Full Product Details